siRNA Details
Gene Symbol: GCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006836.1 | NP_006827.1 | 47 | 6219 | ||
GenBank mRNA | BC040172.1 | - | 3 | 380 | ||
BC153881.1 | - | 47 | 6219 | 8860118 | ||
D86973.2 | - | 47 | 6219 | |||
U77700.1 | - | 25 | 3684 | |||
U88836.1 | - | 8 | 1177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
