siRNA Details
Gene Symbol: GCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006836.1 | NP_006827.1 | 58 | 8019 | ||
GenBank mRNA | BC021129.1 | - | 7 | 891 | ||
BC032335.1 | - | 7 | 846 | |||
BC040172.1 | - | 14 | 2180 | |||
BC046177.1 | - | 4 | 562 | |||
BC064346.1 | - | 6 | 735 | |||
BC113083.1 | - | 3 | 161 | |||
BC113084.1 | - | 3 | 161 | |||
BC153881.1 | - | 58 | 8019 | 8860118 | ||
D86973.2 | - | 58 | 8019 | |||
U77700.1 | - | 36 | 5485 | |||
U88836.1 | - | 19 | 2977 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
