siRNA Details
Gene Symbol: GCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006836.1 | NP_006827.1 | 47, 48 | 6258 | ||
GenBank mRNA | BC040172.1 | - | 3, 4 | 419 | ||
BC153881.1 | - | 47, 48 | 6258 | 8860118 | ||
D86973.2 | - | 47, 48 | 6258 | |||
U77700.1 | - | 25, 26 | 3723 | |||
U88836.1 | - | 8, 9 | 1216 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
