siRNA Details
Gene Symbol: SUDS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022491.2 | NP_071936.2 | 3, 4 | 412 | ||
XM_011538671.2 | XP_011536973.1 | 4, 5 | 1489 | |||
XM_017019817.1 | XP_016875306.1 | 2, 3 | 296 | |||
XM_017019818.1 | XP_016875307.1 | 5, 6 | 644 | |||
XM_017019819.1 | XP_016875308.1 | 4, 5 | 506 | |||
XM_017019820.1 | XP_016875309.1 | 3, 4 | 236 | |||
GenBank mRNA | AK024460.1 | - | 3, 4 | 395 | ||
BC098404.1 | - | 3, 4 | 435 | 30332266 | ||
BX647687.1 | - | 4, 5 | 370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
