siRNA Details
Gene Symbol: DNAJC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287811.1 | NP_001274740.1 | 14 | 5095 | ||
NM_015291.3 | NP_056106.1 | 15 | 5280 | |||
NR_109898.1 | 15 | 5510 | ||||
GenBank mRNA | AB023179.2 | - | 15 | 5236 | ||
AK022530.1 | - | 6 | 3646 | |||
AK023805.1 | - | 11 | 4452 | |||
AK091848.1 | - | 14 | 2763 | |||
AL833299.1 | - | 15 | 5450 | |||
CR749259.1 | - | 14 | 5042 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
