siRNA Details
Gene Symbol: HBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF147332.1 | - | 3 | 524 | ||
BC008572.1 | - | 3 | 529 | 4177243 | ||
V00493.1 | - | 3 | 543 |
Gene Symbol: HBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000517.4 | NP_000508.1 | 3 | 572 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
