siRNA Details
Gene Symbol: SPAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003114.4 | NP_003105.2 | 3 | 565 | ||
NM_172218.2 | NP_757367.1 | 3 | 367 | |||
XM_011517240.2 | XP_011515542.1 | 3 | 367 | |||
XM_011517241.2 | XP_011515543.1 | 3 | 367 | |||
XM_011517242.2 | XP_011515544.1 | 3 | 409 | |||
XM_011517243.2 | XP_011515545.1 | 3 | 303 | |||
XM_011517245.2 | XP_011515547.1 | 3 | 367 | |||
XM_017013754.1 | XP_016869243.1 | 2 | 401 | |||
XM_017013755.1 | XP_016869244.1 | 2 | 354 | |||
GenBank mRNA | AF311312.1 | - | 3 | 479 | ||
AK057482.1 | - | 3 | 367 | |||
BC055091.1 | - | 3 | 346 | |||
GQ472210.1 | - | 3 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
