siRNA Details
Gene Symbol: SPAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003114.4 | NP_003105.2 | 13, 14 | 1969 | ||
NM_172218.2 | NP_757367.1 | 13, 14 | 1771 | |||
XM_011517241.2 | XP_011515543.1 | 13, 14 | 1771 | |||
XM_011517242.2 | XP_011515544.1 | 13, 14 | 1813 | |||
XM_011517243.2 | XP_011515545.1 | 13, 14 | 1707 | |||
XM_017013754.1 | XP_016869243.1 | 12, 13 | 1805 | |||
XM_017013755.1 | XP_016869244.1 | 12, 13 | 1758 | |||
GenBank mRNA | AF311312.1 | - | 13, 14 | 1883 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
