siRNA Details
Gene Symbol: KRT6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005555.3 | NP_005546.2 | 9 | 2004 | ||
GenBank mRNA | BC034535.1 | - | 9 | 2003 | 4754558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
