siRNA Details
Gene Symbol: ALLC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018436.3 | NP_060906.3 | 11 | 1023 | ||
XM_011510369.2 | XP_011508671.1 | 10 | 917 | |||
XM_011510370.2 | XP_011508672.1 | 10 | 3408 | |||
XM_017004495.1 | XP_016859984.1 | 12 | 2102 | |||
XM_017004496.1 | XP_016859985.1 | 9 | 2063 | |||
XM_017004497.1 | XP_016859986.1 | 7 | 650 | |||
XM_017004498.1 | XP_016859987.1 | 5 | 491 | |||
GenBank mRNA | AF215924.1 | - | 4 | 1353 | ||
AF395820.1 | - | 9 | 819 | |||
AK302299.1 | - | 11 | 1023 | |||
BC029652.1 | - | 11 | 999 | 5164086 | ||
BC035356.1 | - | 4 | 1447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
