siRNA Details
Gene Symbol: ALLC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018436.3 | NP_060906.3 | 7 | 643 | ||
XM_011510369.2 | XP_011508671.1 | 6 | 537 | |||
XM_011510370.2 | XP_011508672.1 | 6 | 3028 | |||
XM_017004495.1 | XP_016859984.1 | 8 | 1722 | |||
XM_017004496.1 | XP_016859985.1 | 5 | 1683 | |||
XM_017004497.1 | XP_016859986.1 | 3 | 270 | |||
GenBank mRNA | AF395820.1 | - | 5 | 439 | ||
AK302299.1 | - | 7 | 643 | |||
BC029652.1 | - | 7 | 619 | 5164086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
