siRNA Details
Gene Symbol: ALLC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018436.3 | NP_060906.3 | 3, 4 | 241 | ||
XM_011510369.2 | XP_011508671.1 | 2, 3 | 135 | |||
XM_017004495.1 | XP_016859984.1 | 4, 5 | 1320 | |||
GenBank mRNA | AF395820.1 | - | 1, 2 | 37 | ||
AK302299.1 | - | 3, 4 | 241 | |||
BC029652.1 | - | 3, 4 | 217 | 5164086 | ||
DB054905.1 | - | 3, 4 | 241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
