siRNA Details
Gene Symbol: XPO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015024.4 | NP_055839.3 | 17 | 1987 | ||
XM_017013244.1 | XP_016868733.1 | 17 | 2241 | |||
XM_017013245.1 | XP_016868734.1 | 17 | 2008 | |||
XM_017013246.1 | XP_016868735.1 | 17 | 2210 | |||
XM_017013247.1 | XP_016868736.1 | 17 | 2401 | |||
GenBank mRNA | AB018288.1 | - | 14 | 1351 | ||
AF064729.1 | - | 17 | 1897 | |||
AK291472.1 | - | 17 | 1902 | |||
AK299848.1 | - | 13 | 1423 | |||
BC030785.1 | - | 17 | 1986 | 5269230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
