siRNA Details
Gene Symbol: CCL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201359.1 | NP_001188288.1 | 6 | 623 | ||
NM_005624.3 | NP_005615.2 | 6 | 626 | |||
XM_011528177.2 | XP_011526479.1 | 7 | 827 | |||
XM_017027119.1 | XP_016882608.1 | 7 | 793 | |||
XM_017027120.1 | XP_016882609.1 | 7 | 722 | |||
XM_017027121.1 | XP_016882610.1 | 7 | 668 | |||
XM_017027122.1 | XP_016882611.1 | 7 | 802 | |||
GenBank mRNA | AK292662.1 | - | 6 | 623 | ||
BC130561.1 | - | 5 | 561 | 40146592 | ||
BC144463.1 | - | 5 | 558 | 9052989 | ||
U86358.1 | - | 5 | 515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
