siRNA Details
Gene Symbol: KRTAP9-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031962.2 | NP_114168.1 | 1 | 373 | ||
GenBank mRNA | AJ406947.1 | - | 1 | 373 | ||
BC074918.2 | - | 1 | 365 | 30915341 | ||
BC074919.2 | - | 1 | 365 | 30915560 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
