siRNA Details
Gene Symbol: VIMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203472.2 | NP_982298.2 | 7 | 1021 | ||
GenBank mRNA | BC107774.1 | - | 7 | 951 | 6450503 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
