siRNA Details
Gene Symbol: VIMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018445.5 | NP_060915.2 | 2, 3 | 324 | ||
NM_203472.2 | NP_982298.2 | 2, 3 | 324 | |||
GenBank mRNA | AF157317.1 | - | 2, 3 | 251 | ||
AF328864.1 | - | 2, 3 | 251 | |||
AK026455.1 | - | 2, 3 | 287 | |||
AK097011.1 | - | 1, 2 | 941 | |||
AK225955.1 | - | 2, 3 | 250 | |||
AK314723.1 | - | 2, 3 | 286 | |||
AY324824.1 | - | 2, 3 | 251 | |||
AY618665.1 | - | 2, 3 | 221 | |||
BC005840.2 | - | 2, 3 | 247 | 2967406 | ||
BC107774.1 | - | 2, 3 | 254 | 6450503 | ||
BG829960.1 | - | 2, 3 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
