siRNA Details
Gene Symbol: THAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018105.2 | NP_060575.1 | 3 | 767 | ||
NM_199003.1 | NP_945354.1 | 2 | 571 | |||
GenBank mRNA | AK001339.1 | - | 3 | 761 | ||
AK223231.1 | - | 3 | 746 | |||
AL832077.1 | - | 2 | 545 | |||
BC021721.2 | - | 3 | 741 | 4795229 | ||
CR457256.1 | - | 3 | 530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
