siRNA Details
Gene Symbol: ZNF700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271848.1 | NP_001258777.1 | 2 | 265 | ||
NM_144566.2 | NP_653167.1 | 2 | 256 | |||
GenBank mRNA | AK126871.1 | - | 1 | 477 | ||
AK308305.1 | - | 2 | 242 | |||
AL136732.1 | - | 2 | 239 | |||
BC136777.1 | - | 2 | 183 | 9020767 | ||
BC144406.1 | - | 3 | 304 | |||
DA794204.1 | - | 2 | 256 | |||
DB013169.1 | - | 2 | 223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
