siRNA Details
Gene Symbol: SDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006946.1 | NP_001006947.1 | 6 | 1873 | ||
NM_002997.4 | NP_002988.3 | 5 | 1781 | |||
XM_005262620.4 | XP_005262677.1 | 5 | 1624 | |||
XM_005262621.3 | XP_005262678.3 | 5 | 1504 | |||
XM_005262622.2 | XP_005262679.1 | 5 | 1443 | |||
GenBank mRNA | AJ551176.1 | - | 5 | 1730 | ||
AK222739.1 | - | 4 | 793 | |||
AK299385.1 | - | 5 | 1342 | |||
AK311409.1 | - | 3 | 884 | |||
BC008765.2 | - | 5 | 1725 | 3347793 | ||
J05392.1 | - | 5 | 1683 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
