siRNA Details
Gene Symbol: WDR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015285.2 | NP_056100.2 | 13 | 1836 | ||
NM_052834.2 | NP_443066.2 | 13 | 1836 | |||
XM_006722431.2 | XP_006722494.1 | 13 | 1768 | |||
XM_011525888.1 | XP_011524190.1 | 13 | 1878 | |||
XM_017025682.1 | XP_016881171.1 | 13 | 1741 | |||
XM_017025683.1 | XP_016881172.1 | 3 | 229 | |||
GenBank mRNA | AB011113.2 | - | 13 | 1799 | ||
AF188125.1 | - | 12 | 1625 | |||
AK057141.1 | - | 13 | 1717 | |||
AY099325.1 | - | 12 | 1625 | |||
BC150282.1 | - | 13 | 1799 | 9007268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
