siRNA Details
Gene Symbol: WDR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015285.2 | NP_056100.2 | 28 | 5522 | ||
NM_052834.2 | NP_443066.2 | 27 | 5423 | |||
XM_006722431.2 | XP_006722494.1 | 28 | 5454 | |||
XM_017025682.1 | XP_016881171.1 | 27 | 5328 | |||
XM_017025683.1 | XP_016881172.1 | 18 | 3915 | |||
GenBank mRNA | AB011113.2 | - | 28 | 5489 | ||
BC050352.1 | - | 14 | 2686 | |||
BC150282.1 | - | 28 | 5489 | 9007268 |
Gene Symbol: WDR7-OT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA634713.1 | - | 1 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
