siRNA Details
Gene Symbol: EFR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323553.1 | NP_001310482.1 | 1 | 55 | ||
NM_001323554.1 | NP_001310483.1 | 1 | 55 | |||
NM_001323555.1 | NP_001310484.1 | 1 | 55 | |||
NM_001323558.1 | NP_001310487.1 | 1 | 55 | |||
NM_015137.5 | NP_055952.2 | 1 | 55 | |||
NR_136615.1 | 1 | 55 | ||||
NR_136616.1 | 1 | 55 | ||||
GenBank mRNA | BC071611.1 | - | 1 | 32 | ||
BC108667.1 | - | 1 | 23 | |||
BC152442.1 | - | 1 | 23 | 8862567 | ||
D63477.1 | - | 1 | 23 | |||
DB451954.2 | - | 1 | 34 | |||
DB483230.1 | - | 1 | 55 | |||
DB727938.1 | - | 1 | 34 | |||
HY095205.1 | - | 1 | 51 | |||
HY124760.1 | - | 1 | 24 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
