siRNA Details
Gene Symbol: CLEC10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006344.2 | NP_006335.2 | 9 | 1648 | ||
NM_182906.2 | NP_878910.1 | 9 | 1720 | |||
XM_005256412.1 | XP_005256469.1 | 9 | 1665 | |||
XM_011523613.1 | XP_011521915.1 | 9 | 1755 | |||
XM_011523614.2 | XP_011521916.1 | 10 | 1821 | |||
XM_011523615.1 | XP_011521917.1 | 9 | 1743 | |||
GenBank mRNA | BC039011.1 | - | 9 | 1668 | 5751735 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
