siRNA Details
Gene Symbol: DPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039589.1 | NP_001034678.1 | 5 | 1332 | ||
NM_001319165.1 | NP_001306094.1 | 6 | 2006 | |||
NM_001319166.1 | NP_001306095.1 | 6 | 1743 | |||
NM_001319167.1 | NP_001306096.1 | 6 | 2073 | |||
NM_001319168.1 | NP_001306097.1 | 6 | 1964 | |||
NM_001319169.1 | NP_001306098.1 | 5 | 1792 | |||
NM_001319170.1 | NP_001306099.1 | 6 | 2076 | |||
NM_001319171.1 | NP_001306100.1 | 6 | 2047 | |||
NM_001384.4 | NP_001375.2 | 6 | 2016 | |||
XM_017000502.1 | XP_016855991.1 | 5 | 2098 | |||
XM_017000503.1 | XP_016855992.1 | 5 | 2032 | |||
XM_017000504.1 | XP_016855993.1 | 4 | 2337 | |||
XM_017000505.1 | XP_016855994.1 | 4 | 2269 | |||
XM_017000506.1 | XP_016855995.1 | 5 | 2140 | |||
GenBank mRNA | AK091303.1 | - | 6 | 1935 | ||
BC003181.1 | - | 6 | 1993 | 2958202 | ||
BC016956.1 | - | 6 | 1975 | 3900854 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
