siRNA Details
Gene Symbol: DSTYK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015375.2 | NP_056190.1 | 13 | 3260 | ||
NM_199462.2 | NP_955749.1 | 12 | 3125 | |||
XM_011509392.2 | XP_011507694.1 | 13 | 3191 | |||
XM_011509393.2 | XP_011507695.1 | 14 | 3154 | |||
XM_011509394.2 | XP_011507696.1 | 12 | 2662 | |||
GenBank mRNA | AB007941.1 | - | 8 | 1506 | ||
AY429674.1 | - | 13 | 3232 | |||
BC048204.1 | - | 5 | 1079 | |||
BC053627.1 | - | 12 | 3088 | 6164875 | ||
BC060870.1 | - | 7 | 2234 | 30346187 | ||
BC072406.1 | - | 13 | 3204 | 6138109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
