siRNA Details
Gene Symbol: DSTYK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015375.2 | NP_056190.1 | 13 | 5173 | ||
NM_199462.2 | NP_955749.1 | 12 | 5038 | |||
XM_011509392.2 | XP_011507694.1 | 13 | 5104 | |||
XM_011509393.2 | XP_011507695.1 | 14 | 5067 | |||
XM_011509394.2 | XP_011507696.1 | 12 | 4575 | |||
GenBank mRNA | AB007941.1 | - | 8 | 3420 | ||
AY429674.1 | - | 13 | 5145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
