siRNA Details
Gene Symbol: KL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004795.3 | NP_004786.2 | 4 | 2053 | ||
XM_006719895.2 | XP_006719958.1 | 4 | 1477 | |||
GenBank mRNA | AB005142.1 | - | 4 | 2053 | ||
AK097349.1 | - | 4 | 1529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
