siRNA Details
Gene Symbol: C1QTNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031910.3 | NP_114116.3 | 3 | 2330 | ||
NM_182486.1 | NP_872292.1 | 4 | 1046 | |||
XM_011529857.1 | XP_011528159.1 | 4 | 5187 | |||
XM_017028569.1 | XP_016884058.1 | 3 | 4479 | |||
XM_017028570.1 | XP_016884059.1 | 5 | 3097 | |||
XM_017028571.1 | XP_016884060.1 | 5 | 4835 | |||
XM_017028572.1 | XP_016884061.1 | 5 | 4824 | |||
XM_017028573.1 | XP_016884062.1 | 5 | 2613 | |||
XM_017028574.1 | XP_016884063.1 | 4 | 2406 | |||
GenBank mRNA | AF329842.1 | - | 4 | 984 | ||
AK128125.1 | - | 9 | 4398 | |||
AK226149.1 | - | 2 | 2853 | |||
AY358374.1 | - | 5 | 2467 | |||
BC020551.1 | - | 3 | 2329 | 2900683 | ||
BU542015.1 | - | 1 | 293 | |||
BU859813.1 | - | 1 | 657 | |||
CR456451.1 | - | 4 | 996 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
