siRNA Details
Gene Symbol: IPO8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190995.1 | NP_001177924.1 | 14 | 1700 | ||
NM_006390.3 | NP_006381.2 | 18 | 2338 | |||
XM_017018691.1 | XP_016874180.1 | 18 | 2235 | |||
XM_017018692.1 | XP_016874181.1 | 17 | 1895 | |||
GenBank mRNA | AB209602.1 | - | 5 | 418 | ||
AK302260.1 | - | 14 | 1666 | |||
AU132276.3 | - | 3 | 215 | |||
U77494.1 | - | 18 | 2160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
