siRNA Details
Gene Symbol: IPO8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190995.1 | NP_001177924.1 | 2 | 375 | ||
NM_006390.3 | NP_006381.2 | 6 | 1013 | |||
XM_017018691.1 | XP_016874180.1 | 6 | 910 | |||
XM_017018692.1 | XP_016874181.1 | 5 | 570 | |||
XM_017018693.1 | XP_016874182.1 | 6 | 1006 | |||
GenBank mRNA | AK302260.1 | - | 2 | 341 | ||
BQ435282.1 | - | 6 | 862 | |||
DC400226.1 | - | 2 | 375 | |||
U77494.1 | - | 6 | 835 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
