siRNA Details
Gene Symbol: VHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000551.3 | NP_000542.1 | 3 | 953 | ||
NM_198156.2 | NP_937799.1 | 2 | 830 | |||
GenBank mRNA | AK304002.1 | - | 3 | 538 | ||
AK309560.1 | - | 3 | 877 | |||
BC058831.1 | - | 2 | 681 | 6457060 | ||
L15409.1 | - | 3 | 953 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
