siRNA Details
Gene Symbol: PRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110213.1 | NP_001103683.1 | 1 | 92 | ||
GenBank mRNA | BC095488.1 | - | 1 | 92 | 30327943 | |
BC136499.1 | - | 1 | 83 | 9020488 | ||
BC141916.1 | - | 1 | 91 | |||
BX538206.1 | - | 1 | 93 | |||
BX538207.1 | - | 1 | 93 | |||
BX641094.1 | - | 1 | 96 | |||
BX647085.1 | - | 1 | 14 | |||
BX647155.1 | - | 1 | 92 | |||
BX647156.1 | - | 1 | 86 | |||
BX648436.1 | - | 1 | 93 | |||
BX648656.1 | - | 1 | 22 | |||
BX648901.1 | - | 1 | 22 | |||
CR749583.1 | - | 1 | 90 | |||
K03202.1 | - | 1 | 101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
