siRNA Details
Gene Symbol: PRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110213.1 | NP_001103683.1 | 3 | 163 | ||
GenBank mRNA | BC095488.1 | - | 3 | 163 | 30327943 | |
BC136499.1 | - | 3 | 154 | 9020488 | ||
BC141916.1 | - | 3 | 162 | |||
BX538206.1 | - | 3 | 164 | |||
BX641094.1 | - | 3 | 167 | |||
BX647085.1 | - | 3 | 85 | |||
BX647155.1 | - | 3 | 163 | |||
BX647156.1 | - | 3 | 157 | |||
BX648436.1 | - | 3 | 164 | |||
BX648656.1 | - | 3 | 93 | |||
BX648871.1 | - | 2 | 138 | |||
BX648901.1 | - | 3 | 93 | |||
K03202.1 | - | 3 | 172 |
Gene Symbol: PRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB593029.1 | - | 5 | 499 | ||
BC031043.1 | - | 7 | 728 | |||
BC133676.1 | - | 3 | 167 | 40119187 | ||
BI772091.1 | - | 7 | 699 | |||
K03203.1 | - | 3 | 172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
