siRNA Details
Gene Symbol: PRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110213.1 | NP_001103683.1 | 2, 3 | 140 | ||
GenBank mRNA | BC095488.1 | - | 2, 3 | 140 | 30327943 | |
BC136499.1 | - | 2, 3 | 131 | 9020488 | ||
BC141916.1 | - | 2, 3 | 139 | |||
BX538206.1 | - | 2, 3 | 141 | |||
BX538207.1 | - | 2, 3 | 141 | |||
BX641094.1 | - | 2, 3 | 144 | |||
BX647085.1 | - | 2, 3 | 62 | |||
BX647155.1 | - | 2, 3 | 140 | |||
BX647156.1 | - | 2, 3 | 134 | |||
BX648436.1 | - | 2, 3 | 141 | |||
BX648656.1 | - | 2, 3 | 70 | |||
BX648871.1 | - | 1, 2 | 115 | |||
BX648901.1 | - | 2, 3 | 70 | |||
CR749583.1 | - | 2, 3 | 138 | |||
K03202.1 | - | 2, 3 | 149 |
Gene Symbol: PRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291314.1 | NP_001278243.1 | 5, 6 | 598 | ||
NM_001291315.1 | NP_001278244.1 | 4, 5 | 476 | |||
GenBank mRNA | AB593029.1 | - | 4, 5 | 476 | ||
BC031043.1 | - | 6, 7 | 705 | |||
BC128192.1 | - | 2, 3 | 139 | |||
BC133676.1 | - | 2, 3 | 144 | 40119187 | ||
BI772091.1 | - | 6, 7 | 676 | |||
BM476484.1 | - | 5, 6 | 586 | |||
K03203.1 | - | 2, 3 | 149 |
Gene Symbol: PRH1-PRR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037918.2 | 6, 7 | 704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
