siRNA Details
Gene Symbol: DSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004948.3 | NP_004939.1 | 10 | 1538 | ||
NM_024421.2 | NP_077739.1 | 10 | 1538 | |||
GenBank mRNA | X72925.1 | - | 10 | 1538 | ||
Z34522.1 | - | 10 | 1466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
