siRNA Details
Gene Symbol: GOLM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016548.3 | NP_057632.2 | 10 | 2602 | ||
NM_177937.2 | NP_808800.1 | 10 | 2594 | |||
GenBank mRNA | AF236056.1 | - | 10 | 2544 | ||
AJ420432.1 | - | 1 | 825 | |||
AK027717.1 | - | 10 | 2565 | |||
AK075542.1 | - | 10 | 2537 | |||
AK075557.1 | - | 10 | 2506 | |||
AK092943.1 | - | 11 | 2482 | |||
BC001740.1 | - | 10 | 2517 | 3351316 | ||
EU794616.1 | - | 10 | 2561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
