siRNA Details
Gene Symbol: GOLM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016548.3 | NP_057632.2 | 10 | 1650 | ||
NM_177937.2 | NP_808800.1 | 10 | 1642 | |||
GenBank mRNA | AF236056.1 | - | 10 | 1592 | ||
AK027717.1 | - | 10 | 1613 | |||
AK075542.1 | - | 10 | 1584 | |||
AK075557.1 | - | 10 | 1554 | |||
AK092943.1 | - | 11 | 1530 | |||
BC001740.1 | - | 10 | 1565 | 3351316 | ||
EU794616.1 | - | 10 | 1609 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
