siRNA Details
Gene Symbol: FBXO21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015002.2 | NP_055817.1 | 7 | 971 | ||
NM_033624.2 | NP_296373.1 | 7 | 971 | |||
XM_017019037.1 | XP_016874526.1 | 7 | 976 | |||
XM_017019038.1 | XP_016874527.1 | 7 | 976 | |||
XM_017019039.1 | XP_016874528.1 | 6 | 587 | |||
GenBank mRNA | AB020682.1 | - | 7 | 933 | ||
AF174601.1 | - | 7 | 932 | |||
AK001699.1 | - | 7 | 955 | |||
AK074714.1 | - | 4 | 440 | |||
AK308141.1 | - | 8 | 877 | |||
AK314573.1 | - | 7 | 944 | |||
AL136899.1 | - | 7 | 784 | |||
BC034045.1 | - | 8 | 898 | 4815947 | ||
BC042333.1 | - | 8 | 871 | 5195515 | ||
BC091496.1 | - | 7 | 930 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
