siRNA Details
Gene Symbol: FBXO21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015002.2 | NP_055817.1 | 12 | 3177 | ||
NM_033624.2 | NP_296373.1 | 12 | 3198 | |||
XM_017019037.1 | XP_016874526.1 | 11 | 3023 | |||
XM_017019038.1 | XP_016874527.1 | 11 | 3002 | |||
XM_017019039.1 | XP_016874528.1 | 11 | 2793 | |||
GenBank mRNA | AB020682.1 | - | 12 | 3139 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
