siRNA Details
Gene Symbol: FBXO21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015002.2 | NP_055817.1 | 12 | 1844 | ||
NM_033624.2 | NP_296373.1 | 12 | 1865 | |||
XM_017019037.1 | XP_016874526.1 | 11 | 1690 | |||
XM_017019038.1 | XP_016874527.1 | 11 | 1669 | |||
XM_017019039.1 | XP_016874528.1 | 11 | 1460 | |||
GenBank mRNA | AB020682.1 | - | 12 | 1806 | ||
AF174601.1 | - | 12 | 1805 | |||
AK001699.1 | - | 12 | 1828 | |||
AK308141.1 | - | 13 | 1750 | |||
AK314573.1 | - | 12 | 1817 | |||
AL136899.1 | - | 12 | 1678 | |||
BC034045.1 | - | 13 | 1771 | 4815947 | ||
BC042333.1 | - | 12 | 1564 | 5195515 | ||
BC091496.1 | - | 12 | 1803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
