siRNA Details
Gene Symbol: ELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001422.3 | NP_001413.1 | 3 | 343 | ||
NM_198381.1 | NP_938195.1 | 3 | 475 | |||
XM_017017308.1 | XP_016872797.1 | 3 | 308 | |||
XM_017017309.1 | XP_016872798.1 | 3 | 308 | |||
GenBank mRNA | AF049703.1 | - | 3 | 324 | ||
AF115402.1 | - | 3 | 476 | |||
AF115403.1 | - | 3 | 311 | |||
AK074633.1 | - | 3 | 309 | |||
AK075197.1 | - | 3 | 299 | |||
AK290808.1 | - | 3 | 299 | |||
AK290817.1 | - | 3 | 387 | |||
AK312537.1 | - | 3 | 337 | |||
BC029743.1 | - | 3 | 326 | 4480123 | ||
BG260616.1 | - | 326 | ||||
BP358543.1 | - | 3 | 343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
