siRNA Details
Gene Symbol: ELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243080.1 | NP_001230009.1 | 5 | 1880 | ||
NM_001243081.1 | NP_001230010.1 | 6 | 2063 | |||
NM_001422.3 | NP_001413.1 | 7 | 2165 | |||
NM_198381.1 | NP_938195.1 | 7 | 2297 | |||
XM_017017308.1 | XP_016872797.1 | 6 | 2079 | |||
XM_017017309.1 | XP_016872798.1 | 6 | 2061 | |||
GenBank mRNA | AF115402.1 | - | 7 | 2295 | ||
AF115403.1 | - | 7 | 2130 | |||
AK075197.1 | - | 7 | 2121 | |||
BC029743.1 | - | 7 | 2150 | 4480123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
