siRNA Details
Gene Symbol: CAPRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005898.4 | NP_005889.3 | 3, 4 | 479 | ||
NM_203364.2 | NP_976240.1 | 3, 4 | 479 | |||
XM_017017749.1 | XP_016873238.1 | 2, 3 | 450 | |||
GenBank mRNA | AK127972.1 | - | 2, 3 | 279 | ||
AK299720.1 | - | 2, 3 | 354 | |||
BC001731.1 | - | 3, 4 | 479 | 3355481 | ||
Z48042.1 | - | 2, 3 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
