siRNA Details
Gene Symbol: CAPRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005898.4 | NP_005889.3 | 7 | 927 | ||
NM_203364.2 | NP_976240.1 | 7 | 927 | |||
XM_017017749.1 | XP_016873238.1 | 6 | 898 | |||
GenBank mRNA | AK127972.1 | - | 6 | 727 | ||
BC001731.1 | - | 7 | 927 | 3355481 | ||
Z48042.1 | - | 6 | 780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
