siRNA Details
Gene Symbol: GCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181453.3 | NP_852118.1 | 9 | 3137 | ||
NR_028063.1 | 8 | 3052 | ||||
XM_006712870.3 | XP_006712933.1 | 9 | 3706 | |||
XM_006712871.1 | XP_006712934.1 | 8 | 2960 | |||
XM_011512213.2 | XP_011510515.1 | 8 | 2953 | |||
GenBank mRNA | AB002334.1 | - | 8 | 2933 | ||
BC146789.1 | - | 8 | 2933 | 9007253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
