siRNA Details
Gene Symbol: GPNMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005340.1 | NP_001005340.1 | 1 | 215 | ||
NM_002510.2 | NP_002501.1 | 1 | 215 | |||
XM_005249578.2 | XP_005249635.1 | 1 | 706 | |||
XM_017011676.1 | XP_016867165.1 | 1 | 706 | |||
XM_017011677.1 | XP_016867166.1 | 1 | 706 | |||
XM_017011678.1 | XP_016867167.1 | 1 | 706 | |||
GenBank mRNA | AF322909.1 | - | 1 | 113 | ||
AJ505015.1 | - | 1 | 127 | |||
AK292078.1 | - | 1 | 137 | |||
AK296779.1 | - | 1 | 137 | |||
AK297031.1 | - | 1 | 137 | |||
AK297051.1 | - | 1 | 146 | |||
AK297055.1 | - | 1 | 144 | |||
AK297803.1 | - | 1 | 137 | |||
AK303071.1 | - | 1 | 143 | |||
AK304671.1 | - | 2 | 196 | |||
AK312279.1 | - | 1 | 144 | |||
AY359124.1 | - | 1 | 135 | |||
BC011595.1 | - | 1 | 103 | |||
BC025297.1 | - | 1 | 105 | |||
BC032783.1 | - | 1 | 131 | 5177095 | ||
BM750843.1 | - | 1 | 225 | |||
BT007074.1 | - | 1 | 54 | |||
CD105306.1 | - | 1 | 203 | |||
X76534.1 | - | 1 | 145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
