siRNA Details
Gene Symbol: MMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100600.1 | NP_001094070.1 | 6 | 705 | ||
NR_072989.1 | 7 | 759 | ||||
GenBank mRNA | AK094524.1 | - | 7 | 757 | ||
BC037881.1 | - | 6 | 656 | 5274201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
