siRNA Details
Gene Symbol: IL36B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014438.4 | NP_055253.2 | 3 | 145 | ||
NM_173178.2 | NP_775270.1 | 3 | 145 | |||
XM_011510962.1 | XP_011509264.1 | 3 | 294 | |||
GenBank mRNA | AF200494.1 | - | 3 | 148 | ||
AF201833.1 | - | 3 | 148 | |||
BC101831.1 | - | 2 | 99 | 8069337 | ||
BC101833.1 | - | 2 | 99 | 8069339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
