siRNA Details
Gene Symbol: ORM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000608.2 | NP_000599.1 | 6 | 816 | ||
GenBank mRNA | BC015964.1 | - | 6 | 740 | 3934986 | |
BC056239.1 | - | 6 | 740 | 6463124 |
Gene Symbol: ORM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000607.2 | NP_000598.2 | 6 | 811 | ||
GenBank mRNA | BC026238.1 | - | 6 | 736 | 4767956 | |
M13692.1 | - | 6 | 715 | |||
X02544.1 | - | 6 | 777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
